missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC30A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25352-1-AP-20UL
This item is not returnable.
View return policy
Specifications
TTC30A | |
Polyclonal | |
Unconjugated | |
TTC30A | |
IFT70, TPR repeat protein 30A, TTC30A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100362431, 92104 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q86WT1 | |
TTC30A | |
TTC30A Fusion Protein Ag18361 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TTC30A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction