missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11080-1-AP-20UL
This item is not returnable.
View return policy
Description
LNPEP, also known as insulin-responsive aminopeptidase (IRAP), was initially identified as an abundant protein that is associated with specialized endosomes termed GLUT4 storage vesicles (GSVs) in adipocytes. The luminal domain of LNPEP contains an aminopeptidase activity that is involved in peptide hormone processing and antigenic peptide processing. LNPEP is also a key regulator of GLUT4 trafficking by controlling the sorting of GLUT4 from endosomes to the GSVs.Specifications
UBE2A | |
Polyclonal | |
Unconjugated | |
UBE2A | |
HHR6A, HR6A, RAD6 homolog A, RAD6A, UBC2, UBE2A, Ubiquitin carrier protein A, Ubiquitin protein ligase A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22209, 298317, 7319 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P49459, Q9Z255 | |
UBE2A | |
UBE2A Fusion Protein Ag1567 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
UBE2A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction