missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2R2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14077-1-AP-150UL
This item is not returnable.
View return policy
Description
Protein kinase CK2 is a ubiquitous and pleiotropic Ser/Thr protein kinase involved in cell growth and transformation. This gene encodes a protein similar to the E2 ubiquitin conjugating enzyme UBC3/CDC34. Studies suggest that CK2-dependent phosphorylation of this ubiquitin-conjugating enzyme functions by regulating beta-TrCP substrate recognition and induces its interaction with beta-TrCP, enhancing beta-catenin degradation.Specifications
UBE2R2 | |
Polyclonal | |
Unconjugated | |
UBE2R2 | |
CDC34B, UBC3B, UBE2R2, Ubiquitin carrier protein R2, Ubiquitin protein ligase R2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54926, 67615, 689226 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6ZWZ2, Q712K3 | |
UBE2R2 | |
UBE2R2 Fusion Protein Ag5202 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
UBE2R2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction