Learn More
Fisherbrand™ Vacuum Aspiration Advantage System Accessory, Receiving Bottle
Designed for use with Fisherbrand™ Vacuum Aspiration Advantage Systems.
Brand:  Fisherbrand™ 112523-8
7102.20 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15461934
  • 7380.00 SEK / 4 L
Only null left
Estimated Shipment: 21-08-2025
Add to basket
Description
Description
  • 4L polypropylene receiving bottle
  • For Fisherbrand Aspiration Advantage System
  • Includes lid, in-line filter, tubing and rapid action coupling hose connectors
Fisherbrand Aspiration Advantage System is a compact and portable unit with an integrated resistant diaphragm pump, used for the safe and precise aspiration of non-flammable chemical, biological, and medical liquids.
Fluids can be removed easily and very precisely from slides, petri dishes, microplates, cell culture containers or similar vessels by using different pipettes or glass tips which can be easily connected to the HandVac pipettor attachment.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Aspiration Advantage System
Receiving Bottle
Videos
  • Fisherbrand - Advantage aspiration system
    The Fisherbrand aspiration Advantage system is a compact and portable unit with an integrated resistant diaphragm pump, used for the safe and precise aspiration of non-flammable chemical, biological, and medical liquids.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Fisherbrand Advantage aspiration system
    SDS
    Documents Certificates
    Promotions
    Promotions
    Product Content Correction
    Product Title
    Fisherbrand™ Vacuum Aspiration Advantage System Accessory, Receiving Bottle > Receiving Bottle