missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF654 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24596-1-AP-20UL
This item is not returnable.
View return policy
Description
Zinc finger protein 654 (ZNF654) may be involved in transcriptional regulation.Specifications
ZNF654 | |
Polyclonal | |
Unconjugated | |
ZNF654 | |
Melanoma associated antigen, zinc finger protein 654, ZNF654 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55279, 72020 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8IZM8, Q9DAU9 | |
ZNF654 | |
ZNF654 Fusion Protein Ag20186 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ZNF654 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction