Laboratory Freezers
Laboratory freezers are used to store a variety of samples between −10°C and −30°C, including pharmaceutical, biological, and other commonly used laboratory materials. Products may be upright, freestanding, or under-bench models providing a range of sizes and capacities. An automatic defrost option is available on some products.
Useful Links
Save Now - Exclusive Deals
Product Code 16347191
Product Code 15943620
Product Code 15658416
Product Code 30178702
Complete Your Order - Great Deals
Must Have
Product Code 16347181
Product Code 16337191
Product Code 15668416
FAQ
Regular maintenance is crucial for the optimal performance of -80°C freezers. It is recommended to perform routine checks every three to six months. This includes cleaning the filters, checking the door seals, and ensuring that the temperature control systems are functioning correctly. Annual professional servicing is also advised to address any potential issues and prolong the lifespan of the freezer.
Most -80°C freezers come with built-in battery backup systems that can maintain the temperature for a limited period during power outages. Additionally, many hospital labs invest in external backup generators that can provide power for extended periods. It is essential to regularly test these backup systems to ensure they are operational when needed.
The lifespan of a -80°C freezer can vary depending on the brand, model, and usage conditions. On average, these freezers can last between 10 to 15 years with proper maintenance. Regular servicing and adherence to manufacturer guidelines can help maximize the freezer's operational life, ensuring reliable performance over the years.
By understanding the critical role of ultra-low temperature storage and the key features of -80°C freezers, hospital labs can make informed decisions to enhance their sample preservation capabilities, ultimately supporting better patient outcomes and advancing medical research.