missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADPGK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15639-1-AP-150UL
This item is not returnable.
View return policy
Description
ADPGK.Specifications
ADPGK | |
Polyclonal | |
Unconjugated | |
ADPGK | |
2610017G09Rik, ADP dependent glucokinase, ADP GK, ADPGK, RbBP 35 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
315722, 72141, 83440 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8VDL4, Q9BRR6 | |
Adpgk | |
ADPGK Fusion Protein Ag8105 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ADPGK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction