Learn More
DWK Life Sciences DURAN™ Blank for separating funnel, conical for ISO 4800
For further processing as separating funnel with tap and standard ground joint.
Brand: DWK Life Sciences 210135409
Description
DURAN™ separating funnels are used in chemistry to separate non-mixable liquids. DURAN™ separating funnels Raw bodies are designed for forming the appropriate standard ground joint sleeve according to DIN ISO 4800.
- Conical shape for ISO 4800
- Glass articles for further processing
Specifications
131 mm | |
Borosilicate Glass (3.3) | |
Blank for further processing as dropping funnel; Phase separation |
1000 mL | |
Autoclavable | |
10/Pk. |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.