Learn More
Copper(II)-ethylenediamine complex, technical, 1M sol. in water, not for viscosity determination
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS | 14552-35-3, 7732-18-5 |
Quantity | 1 L |
Molecular Formula | C4H18CuN4O2 |
Linear Formula | Cu(H2NCH2CH2NH2)2(OH)2 |
Flash Point | >110°C |
Solubility Information | Solubility in water: soluble. |
Molecular Weight (g/mol) | 217.76 |
Formula Weight | 217.76 |
Grade | Technical |
Concentration or Composition (by Analyte or Components) | 0.98 to 1.02 M (copper molarity) |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
A0472393 | Certificate of Analysis | 17/05/2025 | 460680010 |
A0462327 | Certificate of Analysis | 25/01/2025 | 460680010 |
A0462327 | Certificate of Analysis | 25/04/2024 | 460680010 |
A0457248 | Certificate of Analysis | 10/04/2024 | 460680010 |
A0365999 | Certificate of Analysis | 21/09/2023 | 460680010 |
Safety and Handling

- Copper(II)-ethylenediamine complex, 1M solution in water
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.