missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIN37 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26651-1-AP-20UL
This item is not returnable.
View return policy
Description
LIN37, also named as Antolefinin and MSTP064, is a component of the DREAM complex (also named LINC complex). LIN37 migrates to 37 kDa for modification.Specifications
LIN37 | |
Polyclonal | |
Unconjugated | |
LIN37 | |
Antolefinin, F25965, lin 37, lin 37 homolog (C. elegans), LIN37, Protein lin 37 homolog, ZK418.4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55957 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96GY3 | |
LIN37 | |
LIN37 Fusion Protein Ag24565 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LIN37 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction