Learn More
Quickfit™ Standard Taper Flat Head Borosilicate Glass Hollow Stopper
Use this glass stopper with Quickfit™ jointed glassware and other glassware having standard ground glass sockets. Azlon™ Standard Taper Flat Head Borosilicate Glass Hollow Stoppers are lightweight, durable and easy to grip.
466.00 SEK - 2185.00 SEK
Specifications
Material | Borosilicate Glass |
---|---|
Product Type | Hollow Stopper |
For Use With (Application) | Chemical durability and high temperature performance |
For Use With (Equipment) | Quickfit™ jointed ware and other Glassware having standard ground Glass sockets |
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | ||||||
12558855
|
Quickfit
SB10 |
Standard; 10/19 |
481.00 SEK
Pack of 5 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
||||||
12508865
|
Quickfit
SB24 |
Standard; 24/29 |
641.00 SEK
Pack of 5 |
In Stock
Log in to see stock. |
||||||
11349623
|
Quickfit
SB34 |
Standard; 34/35 |
1275.00 SEK
Pack of 5 |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
12568855
|
Quickfit
SB19 |
Standard; 19/26 |
477.00 SEK
Pack of 5 |
In Stock
Log in to see stock. |
||||||
12508875
|
Quickfit
SB29 |
Standard; 29/32 |
621.00 SEK
Pack of 5 |
In Stock
Log in to see stock. |
||||||
12495376
|
Quickfit
SB45 |
Standard; 45/40 |
785.00 SEK
Pack of 2 |
Estimated Shipment: 06-08-2025
Log in to see stock. |
||||||
10155824
|
Quickfit
SB14 |
Standard; 14/23 |
466.00 SEK
Pack of 5 |
In Stock
Log in to see stock. |
||||||
15557960
|
Quickfit
SB7 |
7/16 | N/A | |||||||
15537960
|
Quickfit
SB12 |
Standard; 12/21 |
1125.00 SEK
Pack of 10 |
Estimated Shipment: 11-09-2025
Log in to see stock. |
||||||
Description
Use this glass stopper with Quickfit™ jointed glassware and other glassware having standard ground glass sockets. AzlonTM Standard Taper Flat Head Borosilicate Glass Hollow Stoppers are lightweight, durable and easy to grip.
- Conforms to the standard tapers as outlined in ISO 383
- Borosilicate glass for chemical durability and high temperature performance
- Hollow-blown construction to reduce weight
- Hexagonal head prevents the stopper from rolling and provides a good grip
- Top of stopper is flat, allowing them to be stood on end and avoid contamination of the ground surface
Specifications
Borosilicate Glass | |
Hollow Stopper | |
Chemical durability and high temperature performance | |
Quickfit™ jointed ware and other Glassware having standard ground Glass sockets |