missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASSF6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11921-1-AP-20UL
This item is not returnable.
View return policy
Description
RASSF6 is a member of the RAS association domain family of RAS effectors/tumor suppressors that mediate some of the growth inhibitory properties of RAS oncogenes.Specifications
RASSF6 | |
Polyclonal | |
Unconjugated | |
RASSF6 | |
RASSF6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
166824, 305251, 73246 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q4QR82, Q6ZTQ3, Q80UQ2 | |
RASSF6 | |
RASSF6 Fusion Protein Ag2565 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RASSF6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction