Learn More
Vitlab™ Simplex II Bottletop Dispenser
Vitlab™ Simplex II Bottletop Dispenser is designed to be practically universally applicable and can be used with many organic and inorganic solutions.
Brand: Vitlab™ 1621508
5012.60 SEK valid until 2025-07-31
Use promo code "24075" to get your promotional price.
Description
- Direct displacement plunger with broad PFA seal prevents crystallisation build-up
- Settings are quickly and accurately adjusted due to interior scalloped track and practical screw mechanism
- Screwable ventilation plug for easy connection of a drying tube
- Screwable discharge valve is equipped with an additional safety bulb and closes the dosing channel, if dispensing tube is not installed, so that no medium can escape
- Equipped with a positive displacement piston and a fluoroplastic (PFA) sealing lip on the cylinder wall
- Nominal volume 2 to 10mL includes adapters GL 25, GL 28, GL 32, GL 38, S 40 and telescopic intake tube (length 125 to 240mm)
- Nominal volume 25 to 100mL includes adapters GL 32, GL 38, S 40 and telescopic intake tube (length 170 to 330mm)
Specifications
10.0 to 100.0 mL |
Product Suggestions
Customers who viewed this item also viewed.
Promotions
-
Multiple Manufacturers
The Ultimate Lab Appliances Promotion
Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.Promo Code: 24075Expires: 2025-07-31
Days
8Hours
19Minutes
48Seconds
28