missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STT3A Mouse anti-Human, Mouse, Clone: 1E2B12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66581-1-IG-20UL
This item is not returnable.
View return policy
Description
This gene encodes the basic transcription factor 3. This protein forms a stable complex with RNA polymerase IIB and is required for transcriptional initiation. Alternative splicing results in multiple transcript variants encoding different isoforms. This gene has multiple pseudogenes.Specifications
STT3A | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P46977, P46978 | |
STT3A | |
STT3A Fusion Protein Ag27072 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1E2B12 | |
Unconjugated | |
STT3A | |
B5, Integral membrane protein 1, ITM1, STT3 A, STT3A, TMC, Transmembrane protein TMC | |
Mouse | |
Protein G | |
RUO | |
16430, 3703 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
STT3A Mouse anti-Human, Mouse, Clone: 1E2B12, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction