CAS RN 117-81-7
CAS RN 117-81-7
IUPAC Name:
1,2-bis(2-ethylhexyl) benzene-1,2-dicarboxylate
Synonyms:
etalon ergoplast fdo-S sansocizer R 8000 flexol dop diacizer dop di-sec-octyl phthalate kodaflex dehp bis(ethylhexyl) phthalate phthalate, diethylhexyl hercoflex 260
Molecular Weight (g/mol):
390.56
Molecular Formula:
C24H38O4
InChi Key:
BJQHLKABXJIVAM-UHFFFAOYNA-N
SMILES:
CCCCC(CC)COC(=O)C1=CC=CC=C1C(=O)OCC(CC)CCCC