Learn More
Gilac™ Two-tone Nesting Stacking Container
Two-tone Nesting Stacking Container 600 x 400 is highly durable in the intensive use cycle and easy cleaning. Benefits from ergonomic integrated handles for an easy and practical grip.
Brand: Gilac™ G180702
This item is not returnable.
View return policy
Description
- Stacking and nesting solid container.
- Highly durable in the intensive use cycle.
- Ergonomic integrated handles for an easy and practical grip.
- Smooth sides and bottom for easy cleeaning.
- Ideal container in market gardening for harvesting and transporting fruit and vegetables.
- Stacking and nesting when half turned.
- Quick recognition thanks to the two-tone system.
- If the containers are full and stacked, all colors are on the same side.
- If they are empty and nested, the colors are alternated.
- 1 embossed labeling area for easy label removal.
- Compatible with 4 and 6-Wheel GILAC rolling dollies.
Specifications
9 gal. | |
None | |
HDPE | |
Stacking Container | |
600 x 400 x 184 mm | |
23.6 in. | |
15.7 in. | |
7.2 in. |
34 L | |
Blue, Red | |
Rectangular | |
Yes | |
600 mm | |
400 mm | |
184 mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Gilac™ Two-tone Nesting Stacking Container > Dimensions (L x W x H): 600 x 400 x 184 mm, Capacity: 34 L, Color: Blue, Red
Spot an opportunity for improvement?Share a Content Correction