Learn More
BRAND™ Amber 3.3 Borosilicate Glass Volumetric Flask With Polypropylene Stoppers
Amber glass to protect light-sensitive contents.
Brand:  BRAND™ 37491
1040.25 SEK valid until 2025-12-31
Use promo code "24375" to get your promotional price.
 View more versions of this product
Product Code. 15693967
  • 1095.00 SEK / Each
Only null left
Estimated Shipment: 04-07-2025
Description
  • Manufactured from Boro 3.3 glass
  • Acc. to DIN EN ISO 1042
  • Includes USP batch certificate
  • Class A, DE-M marking
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PP (Polypropylene)
Borosilicate Glass (3.3)
A
1000 mL
Volumetric flask
Stopper
Videos
  • Brand - BLAUBRAND
    BLAUBRAND video
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Brand BLAUBRAND
    SDS
    Documents Certificates
    Promotions
    Promotions
    Product Content Correction
    Product Title
    BRAND™ Amber 3.3 Borosilicate Glass Volumetric Flask With Polypropylene Stoppers > Capacity (Metric): 1000 mL