CAS RN 1134-47-0
CAS RN 1134-47-0
IUPAC Name:
4-amino-3-(4-chlorophenyl)butanoic acid
Synonyms:
baclofen nu baclo novartis brand of baclofen lioresal intrathecal medtronic brand of baclofen NuBaclo baclofen awd baclofen alphapharm brand baclofene baclofeno
Molecular Weight (g/mol):
213.66
Molecular Formula:
C10H12ClNO2
InChi Key:
KPYSYYIEGFHWSV-UHFFFAOYNA-N
SMILES:
NCC(CC(O)=O)C1=CC=C(Cl)C=C1
1
–
3
of
3
results
3
Medchem Express Baclofen, MedChemExpress
MedChemExpress Baclofen, a lipophilic derivative of γ-aminobutyric acid (GABA), is an orally active, selective metabotropic GABAB receptor (GABABR) agonist. Baclofen mimics the action of GABA and produces slow presynaptic inhibition through the GABAB receptor. Baclofen has high blood brain barrier penetrance. Baclofen has the potential for muscle spasticity research.
