Learn More
Quickfit™ Borosilicate Glass Single Neck Pear Shaped Flask
Allows distillation down to small quantities.
Brand: Quickfit FP50/2
This item is not returnable.
View return policy
Description
Allows distillation down to small quantities.
- With highest quality Quickfit™ ground glass socket
- Ideal for semi-micro distillations
- Pear shaped design allows distillation down to small quantities with minimum hold-up, and reduced losses from thermal decomposition
- Borosilicate glass for chemical resistance and thermal performance
Specifications
Pear | |
Borosilicate Glass | |
48 mm |
50 mL | |
125 mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product information
File size: 2.8 MB
Product Content Correction
Product Title
Quickfit™ Borosilicate Glass Single Neck Pear Shaped Flask > Capacity: 50mL; Socket: 19/26
Spot an opportunity for improvement?Share a Content Correction